Merck
Human Cytomegalovirus Compromises Development of Cerebral Organoids.
Journal of virology (2019-06-21)
Rebecca M Brown, Pranav S J B Rana, Hannah K Jaeger, John M O'Dowd, Onesmo B Balemba, Elizabeth A Fortunato
ABSTRACT
Congenital human cytomegalovirus (HCMV) infection causes a broad spectrum of central and peripheral nervous system disorders, ranging from microcephaly to hearing loss. These ramifications mandate the study of virus-host interactions in neural cells. Neural progenitor cells are permissive for lytic infection. We infected two induced pluripotent stem cell (iPSC) lines and found these more primitive cells to be susceptible to infection but not permissive. Differentiation of infected iPSCs induced de novo expression of viral antigens. iPSCs can be cultured in three dimensions to generate cerebral organoids, closely mimicking in vivo development. Mock- or HCMV-infected iPSCs were subjected to a cerebral organoid generation protocol. HCMV IE1 protein was detected in virus-infected organoids at 52 days postinfection. Absent a significant effect on organoid size, infection induced regions of necrosis and the presence of large vacuoles and cysts. Perhaps more in parallel with the subtler manifestations of HCMV-induced birth defects, infection dramatically altered neurological development of organoids, decreasing the number of developing and fully formed cortical structure sites, with associated changes in the architectural organization and depth of lamination within these structures, and manifesting aberrant expression of the neural marker β-tubulin III. Our observations parallel published descriptions of infected clinical samples, which often contain only sparse antigen-positive foci yet display areas of focal necrosis and cellular loss, delayed maturation, and abnormal cortical lamination. The parallels between pathologies present in clinical specimens and the highly tractable three-dimensional (3D) organoid system demonstrate the utility of this system in modeling host-virus interactions and HCMV-induced birth defects.IMPORTANCE Human cytomegalovirus (HCMV) is a leading cause of central nervous system birth defects, ranging from microcephaly to hearing impairment. Recent literature has provided descriptions of delayed and abnormal maturation of developing cortical tissue in infected clinical specimens. We have found that infected induced pluripotent stem cells can be differentiated into three-dimensional, viral protein-expressing cerebral organoids. Virus-infected organoids displayed dramatic alterations in development compared to those of mock-infected controls. Development in these organoids closely paralleled observations in HCMV-infected clinical samples. Infection induced regions of necrosis, the presence of larger vacuoles and cysts, changes in the architectural organization of cortical structures, aberrant expression of the neural marker β-tubulin III, and an overall reduction in numbers of cortical structure sites. We found clear parallels between the pathologies of clinical specimens and virus-infected organoids, demonstrating the utility of this highly tractable system for future investigations of HCMV-induced birth defects.
Sigma-Aldrich
Anti-Nestin Antibody, clone 10C2, clone 10C2, Chemicon®, from mouse