Merck
Biodegradable Thermomagnetically Responsive Soft Untethered Grippers.
ACS applied materials & interfaces (2018-12-12)
Kunihiko Kobayashi, ChangKyu Yoon, Seung Hyun Oh, Jayson V Pagaduan, David H Gracias
ABSTRACT
Soft-robotic devices such as polymeric microgrippers offer the possibility for pick and place of fragile biological cargo in hard-to-reach conduits with potential applications in drug delivery, minimally invasive surgery, and biomedical engineering. Previously, millimeter-sized self-folding thermomagnetically responsive soft grippers have been designed, fabricated, and utilized for pick-and-place applications but there is a concern that such devices could get lost or left behind after their utilization in practical clinical applications in the human body. Consequently, strategies need to be developed to ensure that these soft-robotic devices are biodegradable so that they would disintegrate if left behind in the body. In this paper, we describe the photopatterning of bilayer gels composed of a thermally responsive high-swelling poly(oligoethylene glycol methyl ether methacrylate ( Mn = 500)-bis(2-methacryloyl)oxyethyl disulfide), P(OEGMA-DSDMA), and a low-swelling poly(acrylamide- N, N'-bis(acyloyl)cystamine) hydrogel, in the shape of untethered grippers. These grippers can change shape in response to thermal cues and open and close due to the temperature-induced swelling of the P(OEGMA-DSDMA) layer. We demonstrate that the grippers can be doped with magnetic nanoparticles so that they can be moved using magnetic fields or loaded with chemicals for potential applications as drug-eluting theragrippers. Importantly, they are also biodegradable at physiological body temperature (∼37 °C) on the basis of cleavage of disulfide bonds by reduction. This approach that combines thermoresponsive shape change, magnetic guidance, and biodegradability represents a significant advance to the safe implementation of untethered shape-changing biomedical devices and soft robots for medical and surgical applications.