Merck
  • Exosomes increased angiogenesis in papillary thyroid cancer microenvironment.
Exosomes increased angiogenesis in papillary thyroid cancer microenvironment.
Endocrine-related cancer (2019-03-15)
Feng Wu, Fuxingzi Li, Xiao Lin, Feng Xu, Rongrong Cui, Jiayu Zhong, Ting Zhu, Su-Kang Shan, Xiao-Bo Liao, Lingqing Yuan, Zhaohui Mo
ABSTRACT
Tumour-derived exosomes under hypoxic conditions contain informative microRNAs involved in the interaction of cancer and para-carcinoma cells, thus contributing to tissue remodelling of the tumour microenvironment (TME). Exosomes isolated from hypoxic papillary thyroid cancer cells, BCPAP cells and KTC-1 cells, enhanced the angiogenesis of human umbilical vein endothelial cells (HUVECs) compared with exosomes isolated from normal thyroid follicular cell line (Nthy-ori-3-1), normoxic BCPAP cells or KTC-1 cells both in vitro and vivo. miR-21-5p was significantly upregulated in exosomes from papillary thyroid cancer BCPAP cells under hypoxic conditions, while the exosomes isolated from hypoxic BCPAP cells with knock-down of miR-21-5p attenuated the promoting effect of angiogenesis. In addition, our experiment revealed that miR-21-5p directly targeted and suppressed TGFBI and COL4A1, thereby increasing endothelial tube formation. Furthermore, elevated levels of exosomal miR-21-5p are found in the sera of papillary thyroid cancer patients, which promote the angiogenesis of HUVECs. Taken together, our study reveals the cell interaction between hypoxic papillary thyroid cancer cells and endothelial cells, elucidating a new mechanism by which hypoxic papillary thyroid cancer cells increase angiogenesis via exosomal miR-21-5p/TGFBI and miR-21-5p/COL4A1 regulatory pathway.