Merck
  • Exploiting interconnected synthetic lethal interactions between PARP inhibition and cancer cell reversible senescence.
Exploiting interconnected synthetic lethal interactions between PARP inhibition and cancer cell reversible senescence.
Nature communications (2019-06-13)
Hubert Fleury, Nicolas Malaquin, Véronique Tu, Sophie Gilbert, Aurélie Martinez, Marc-Alexandre Olivier, Alexandre Sauriol, Laudine Communal, Kim Leclerc-Desaulniers, Euridice Carmona, Diane Provencher, Anne-Marie Mes-Masson, Francis Rodier
ABSTRACT
Senescence is a tumor suppression mechanism defined by stable proliferation arrest. Here we demonstrate that the known synthetic lethal interaction between poly(ADP-ribose) polymerase 1 inhibitors (PARPi) and DNA repair triggers p53-independent ovarian cancer cell senescence defined by senescence-associated phenotypic hallmarks including DNA-SCARS, inflammatory secretome, Bcl-XL-mediated apoptosis resistance, and proliferation restriction via Chk2 and p21 (CDKN1A). The concept of senescence as irreversible remains controversial and here we show that PARPi-senescent cells re-initiate proliferation upon drug withdrawal, potentially explaining the requirement for sustained PARPi therapy in the clinic. Importantly, PARPi-induced senescence renders ovarian and breast cancer cells transiently susceptible to second-phase synthetic lethal approaches targeting the senescence state using senolytic drugs. The combination of PARPi and a senolytic is effective in preclinical models of ovarian and breast cancer suggesting that coupling these synthetic lethalities provides a rational approach to their clinical use and may together be more effective in limiting resistance.