- Transcriptomic analysis of Campylobacter jejuni grown in a medium containing serine as the main energy source.
Transcriptomic analysis of Campylobacter jejuni grown in a medium containing serine as the main energy source.
Campylobacter jejuni is one of the most important causes of food-borne diseases in industrialized countries. Amino acids are an important nutrient source for this pathogen because it lacks enzymes related to glycolysis. However, the metabolic characteristics of C. jejuni grown in a nutrient-restricted medium with specific amino acids have not been fully elucidated. This study shows that C. jejuni NCTC 11168 grows well in a nutrient-restricted medium containing serine, aspartate, glutamate, and proline. Subtracting serine significantly reduced growth, but the removal of the three other amino acids did not, suggesting that serine is a priority among the four amino acids. A transcriptomic analysis of C. jejuni NCTC 11168 grown in a medium with serine as the main energy source was then performed. Serine seemed to be sensed by some chemoreceptors, and C. jejuni reached an adaptation stage with active growth in which the expression of flagellar assembly components was downregulated and the biosyntheses of multiple amino acids and nucleotide sugars were upregulated. These data suggest that C. jejuni NCTC 11168 requires serine as a nutrient.