Merck
  • A regio- and stereoselective synthesis of trisubstituted alkenes via gold(I)-catalyzed hydrophosphoryloxylation of haloalkynes.
A regio- and stereoselective synthesis of trisubstituted alkenes via gold(I)-catalyzed hydrophosphoryloxylation of haloalkynes.
Chemical communications (Cambridge, England) (2011-06-07)
Bathoju Chandra Chary, Sunggak Kim, Doosup Shin, Phil Ho Lee
ABSTRACT
A new stereoselective synthesis of trisubstituted alkenes is developed. Hydrophosphoryloxylation of haloalkynes provides Z-alkenyl halophosphates, which undergo Pd-catalyzed consecutive cross-coupling reactions to afford regio- and stereodefined trisubstituted alkenes.
Sigma-Aldrich
Vinylboronic acid pinacol ester, contains phenothiazine as stabilizer, 95%