Merck
  • Management of Developmental Dysplasia of the Hip in Infants up to Six Months of Age: Intended for Use by General Pediatricians and Referring Physicians.
Management of Developmental Dysplasia of the Hip in Infants up to Six Months of Age: Intended for Use by General Pediatricians and Referring Physicians.
The Journal of the American Academy of Orthopaedic Surgeons (2018-10-10)
Samora Julie, Robert H Quinn, Jayson Murray, Ryan Pezold, Quinn Hall
ABSTRACT
The American Academy of Orthopaedic Surgeons has developed the Appropriate Use Criteria (AUC) for the Management of Developmental Dysplasia of the Hip in Infants up to Six Months of Age: Intended for Use by General Pediatricians and Referring Physicians. Evidence-based information, in conjunction with the clinical expertise of physicians, was used to develop the criteria to improve patient care and obtain best outcomes while considering the subtleties and distinctions necessary in making clinical decisions. The Management of Developmental Dysplasia of the Hip in Infants up to Six Months of Age: Intended for Use by General Pediatricians and Referring Physicians AUC clinical patient scenarios were derived from indications typical of patients presenting with developmental dysplasia of the hip in clinical practice. The 24 patient scenarios and four treatments were developed by the Writing Panel, a group of clinicians who are specialists in this AUC topic. Next, a separate multidisciplinary, Voting Panel (made up of specialists and nonspecialists) rated the appropriateness of treatment of each patient scenario using a 9-point scale to designate a treatment as Appropriate (median rating, 7 to 9), May be Appropriate (median rating, 4 to 6), or Rarely Appropriate (median rating, 1 to 3).