- Pd(II)-Catalyzed Enantioselective C(sp3)-H Arylation of Free Carboxylic Acids.
Pd(II)-Catalyzed Enantioselective C(sp3)-H Arylation of Free Carboxylic Acids.
Journal of the American Chemical Society (2018-05-10)
Peng-Xiang Shen, Liang Hu, Qian Shao, Kai Hong, Jin-Quan Yu
PMID29741883
ABSTRACT
A monoprotected aminoethyl amine chiral ligand based on an ethylenediamine backbone was developed to achieve Pd-catalyzed enantioselective C(sp3)-H arylation of cyclopropanecarboxylic and 2-aminoisobutyric acids without using exogenous directing groups. This new chiral catalyst affords new disconnection for preparing diverse chiral carboxylic acids from simple starting materials that are complementary to the various ring forming approaches.
Product Description
(S)-N-(1-(Dimethylamino)-3-phenylpropan-2-yl)acetamide, ≥95%