- Cu-mediated chemoselective trifluoromethylation of benzyl bromides using shelf-stable electrophilic trifluoromethylating reagents.
Cu-mediated chemoselective trifluoromethylation of benzyl bromides using shelf-stable electrophilic trifluoromethylating reagents.
Organic letters (2011-06-15)
Hiroyuki Kawai, Tatsuya Furukawa, Yoshinori Nomura, Etsuko Tokunaga, Norio Shibata
PMID21667988
ABSTRACT
Copper-mediated chemoselective trifluoromethylation at the benzylic position by the use of shelf-stable electrophilic trifluoromethylating reagents 3 in good to high yields under mild conditions is described for the first time. The generality of this trifluoromethylation for a wide variety of benzyl bromides facilitates the rapid creation of structural diversity of medicinal candidates in drug discovery.