Merck
  • Analysis of the interactions of viral and cellular factors with human cytomegalovirus lytic origin of replication, oriLyt.
Analysis of the interactions of viral and cellular factors with human cytomegalovirus lytic origin of replication, oriLyt.
Virology (2012-01-13)
Dominique Kagele, Cyprian C Rossetto, Margaret T Tarrant, Gregory S Pari
ABSTRACT
Human cytomegalovirus transient lytic DNA replication relies on the cis-acting element oriLyt, six viral-encoded core proteins, the proposed DNA replication initiator protein UL84, IE2, IRS1 and the gene products from the UL112/113 loci. In an effort to elucidate cellular and viral-encoded factors that may play a role in oriLyt-dependent replication we used DNA-affinity purification and mass spectrometry to isolate and identify several previously unknown cellular and viral factors that interact with HCMV oriLyt DNA. These proteins include the multifunctional hnRNP-K, BUB3, HMGB1, PTB-1, UL83, UL112/113, and IRS1. Chromatin immunoprecipitation (ChIP) assays confirmed an interaction of several of these factors with oriLyt. Co-immunoprecipitation experiments detected an interaction between UL84 and hnRNP-K in infected and transfected cells. Knockdown of hnRNP K expression by siRNA inhibited the amplification of oriLyt in the transient assay. Together, these data suggest a possible regulatory role in DNA replication for several previously unidentified viral and cellular factors.