Merck
  • Benzoxazole benzenesulfonamides as allosteric inhibitors of fructose-1,6-bisphosphatase.
Benzoxazole benzenesulfonamides as allosteric inhibitors of fructose-1,6-bisphosphatase.
Bioorganic & medicinal chemistry letters (2006-02-01)
Chunqiu Lai, Rebecca J Gum, Melissa Daly, Elizabeth H Fry, Charles Hutchins, Celerino Abad-Zapatero, Thomas W von Geldern
ABSTRACT
A series of novel benzoxazole benzenesulfonamides was synthesized as inhibitors of fructose-1,6-bisphosphatase (FBPase-1). Extensive SAR studies led to a potent inhibitor, 53, with an IC(50) of 0.57microM. Compound 17 exhibited excellent bioavailability and a good pharmacokinetic profile in rats.
Sigma-Aldrich
2,5-Dichlorobenzenesulfonyl chloride, 98%