Merck
  • Interferon-stimulated gene 60 (ISG60) constitutes a negative feedback loop in the downstream of TLR3 signaling in hCMEC/D3 cells.
Interferon-stimulated gene 60 (ISG60) constitutes a negative feedback loop in the downstream of TLR3 signaling in hCMEC/D3 cells.
Journal of neuroimmunology (2018-09-10)
Tadaatsu Imaizumi, Naoko Sassa, Shogo Kawaguchi, Tomoh Matsumiya, Hidemi Yoshida, Kazuhiko Seya, Toshihiro Shiratori, Koji Hirono, Hiroshi Tanaka
ABSTRACT
Brain capillary endothelial cells are the component of blood brain barrier, and the first line of defense against viruses invading into brain. We demonstrate that treatment of hCMEC/D3 cells, a human brain capillary endothelial cell line, with a Toll-like receptor 3 (TLR3) agonist polyinosinic-polycytidylic acid (poly IC) induces the expression of interferon (IFN)-stimulated gene 60 (ISG60), and this reaction was mediated by IFN-β. Knockdown of ISG60 increased the poly IC-induced expression of IFN-β and an IFN-β-inducible chemokine CXCL10. This indicates that ISG60 constitutes a negative feedback loop in the downstream of TLR3/IFN-β. ISG60 in brain capillary endothelial cells may contribute to prevent excess immune reactions associated with viral infections.
Sigma-Aldrich
MISSION® esiRNA, targeting human TLR3