Merck
  • Discovery of BI-2545: A Novel Autotaxin Inhibitor That Significantly Reduces LPA Levels in Vivo.
Discovery of BI-2545: A Novel Autotaxin Inhibitor That Significantly Reduces LPA Levels in Vivo.
ACS medicinal chemistry letters (2017-12-21)
Christian A Kuttruff, Marco Ferrara, Tom Bretschneider, Stefan Hoerer, Sandra Handschuh, Bernd Nosse, Helmut Romig, Paul Nicklin, Gerald J Roth
ABSTRACT
In an effort to find new therapeutic interventions addressing the unmet medical need of patients with idiopathic pulmonary fibrosis, we initiated a program to identify new autotaxin (ATX) inhibitors. Starting from a recently published compound (PF-8380), we identified several highly potent ATX inhibitors with improved pharmacokinetic and safety profiles. Further optimization efforts resulted in the identification of a single-digit nanomolar lead compound (BI-2545) that shows substantial lowering of LPA in vivo and is therefore considered a valuable tool for further studies.
Sigma-Aldrich
BI-2545, ≥98% (HPLC)