Merck
  • Glucocerebrosidase haploinsufficiency in A53T α-synuclein mice impacts disease onset and course.
Glucocerebrosidase haploinsufficiency in A53T α-synuclein mice impacts disease onset and course.
Molecular genetics and metabolism (2017-11-28)
Nahid Tayebi, Loukia Parisiadou, Bahafta Berhe, Ashley N Gonzalez, Jenny Serra-Vinardell, Raphael J Tamargo, Emerson Maniwang, Zachary Sorrentino, Hideji Fujiwara, Richard J Grey, Shahzeb Hassan, Yotam N Blech-Hermoni, Chuyu Chen, Ryan McGlinchey, Chrissy Makariou-Pikis, Mieu Brooks, Edward I Ginns, Daniel S Ory, Benoit I Giasson, Ellen Sidransky
ABSTRACT
Mutations in GBA1 encountered in Gaucher disease are a leading risk factor for Parkinson disease and associated Lewy body disorders. Many GBA1 mutation carriers, especially those with severe or null GBA1 alleles, have earlier and more progressive parkinsonism. To model the effect of partial glucocerebrosidase deficiency on neurological progression in vivo, mice with a human A53T α-synuclein (SNCAA53T) transgene were crossed with heterozygous null gba mice (gba+/-). Survival analysis of 84 mice showed that in gba+/-//SNCAA53T hemizygotes and homozygotes, the symptom onset was significantly earlier than in gba+/+//SNCAA53T mice (p-values 0.023-0.0030), with exacerbated disease progression (p-value <0.0001). Over-expression of SNCAA53T had no effect on glucocerebrosidase levels or activity. Immunoblotting demonstrated that gba haploinsufficiency did not lead to increased levels of either monomeric SNCA or insoluble high molecular weight SNCA in this model. Immunohistochemical analyses demonstrated that the abundance and distribution of SNCA pathology was also unaltered by gba haploinsufficiency. Thus, while the underlying mechanism is not clear, this model shows that gba deficiency impacts the age of onset and disease duration in aged SNCAA53T mice, providing a valuable resource to identify modifiers, pathways and possible moonlighting roles of glucocerebrosidase in Parkinson pathogenesis.