Merck
  • The novel multifunctional, iron-chelating drugs M30 and HLA20 protect pancreatic beta-cell lines from oxidative stress damage.
The novel multifunctional, iron-chelating drugs M30 and HLA20 protect pancreatic beta-cell lines from oxidative stress damage.
The Journal of pharmacology and experimental therapeutics (2010-03-20)
Danit Mechlovich, Tamar Amit, Silvia A Mandel, Orit Bar-Am, Konstantin Bloch, Pnina Vardi, Moussa B H Youdim
ABSTRACT
Increasing evidence suggests that oxidative stress (OS)-induced pancreatic beta-cell impairments is involved in diabetes and diabetic complications. Our group has recently synthesized two multifunctional nontoxic, lipophilic, iron-chelating drugs, 5-{N-methyl-N-propargylaminomethyl}-8-hydroxyquinoline (M30) and 5-{4-propargylpiperazin-1-ylmethyl}-8-hydroxyquinoline (HLA20), for the treatment of various OS-mediated pathogeneses. These compounds contain the N-propargylamine cytoprotective moiety of the antiparkinsonian drug rasagiline (Azilect) and the iron-complexing component 8-hydroxyquinoline. The aim of this research was to evaluate the protective effect of the multifunctional iron-chelating drugs on rat insulin-producing pancreatic beta-cells (INS-1E and RINm) against OS-induced cytotoxicity. We found that M30 and HLA20 markedly and dose-dependently inhibited H(2)O(2)-induced cytotoxicity, associated with decreased intracellular reactive oxygen species formation and increased catalase activity. In accordance, the catalase inhibitor 3-amino-1,2,4-triazol blocked the protective action of M30 against H(2)O(2)-induced damage. Both compounds significantly increased the levels of the iron-responsive protein transferrin receptor indicating their iron-chelating effect. Further mechanistic studies showed that M30 and HLA20 attenuated H(2)O(2)-induced mitochondrial membrane potential loss, decreased the release of cytochrome c into the cytoplasm, and inhibited the activation of caspase-3, suggesting that these drugs may produce cytoprotective effects via the preservation of mitochondrial function. These results indicate that the novel drugs, M30 and HLA20 display significant cytoprotective activity against OS-induced cytotoxicity in insulin producing beta-cells, which might be of therapeutic use in the treatment of diabetes mellitus.