Merck
  • Proton Affinitive Derivatization for Highly Sensitive Determination of Testosterone and Dihydrotestosterone in Saliva Samples by LC-ESI-MS/MS.
Proton Affinitive Derivatization for Highly Sensitive Determination of Testosterone and Dihydrotestosterone in Saliva Samples by LC-ESI-MS/MS.
Analytical sciences : the international journal of the Japan Society for Analytical Chemistry (2018-08-07)
Ken-Ichi Ohno, Tomomi Hasegawa, Tomomi Tamura, Haruka Utsumi, Kouwa Yamashita
ABSTRACT
In this study, proton affinitive derivatization using picolinic acid and its analogs (3- and 6-methylpicolinic acid and 5-butylpicolinic acid) with proton affinitive moieties was performed for the highly sensitive determination of testosterone (T) and 5α-dihydrotestosterone (DHT) in saliva by LC-ESI-MS/MS. T and DHT were converted to their corresponding picolinate esters and their chromatographic behavior was investigated with a reversed phase column. The picolinate ester of each steroid exhibited a clear single peak and elution occurred in the following order: picolinate, 3/6-methylpicolinate, and 5-butylpicolinate. Estimation and understanding of the separation and retention time of each picolinate ester was made simple using the develop method. Although the peaks of picolinate and 3/6-methylpicolinate esters were suppressed by interference from the saliva background (matrix effect), the 5-butylpicolinate esters were only marginally affected.
Sigma-Aldrich
2-Methyl-6-nitrobenzoic anhydride, 97%
Sigma-Aldrich
3-Methylpyridine-2-carboxylic acid, 97%