Merck
  • The O-methylated isoflavone, formononetin, inhibits human ovarian cancer cell proliferation by sub G0/G1 cell phase arrest through PI3K/AKT and ERK1/2 inactivation.
The O-methylated isoflavone, formononetin, inhibits human ovarian cancer cell proliferation by sub G0/G1 cell phase arrest through PI3K/AKT and ERK1/2 inactivation.
Journal of cellular biochemistry (2018-05-16)
Sunwoo Park, Fuller W Bazer, Whasun Lim, Gwonhwa Song
ABSTRACT
Formononetin is an isoflavone that is extracted from red clovers or soy. It has anti-oxidant, anti-proliferative, and anti-tumor effects against cells in various diseases. Several cohort studies have indicated that phytoestrogen intake, including formononetin, could reduce the risk of various carcinogenesis. In fact, many case-control studies have indicated the potential value of flavonoids as drug supplements in the treatment of many cancer patients. However, the toxic effects and the anti-cancer mechanism of formononetin in ovarian cancer are unknown. We investigated the toxicological mechanism of formononetin in ES2 and OV90 ovarian cancer cells. Formononetin suppressed cell proliferation through sub G0/G1 phase arrest and increased apoptosis in both cell lines. Furthermore, it induced loss of mitochondrial membrane potential and generation of reactive oxygen species in ES2 and OV90 cells. The formononetin-mediated regulation of cell proliferation and apoptosis involved decreased phosphorylation of ERK1/2, P90RSK, AKT, P70S6K, and S6 proteins, and increased phosphorylation of P38 protein in ES2 and OV90 cells. Co-treatment of formononetin with pharmacological inhibitors (LY294002 or U0126) revealed additional anti-proliferative effects on the two human ovarian cancer cell types. Conclusively, the results indicate the potential value of formononetin as an anti-cancer agent in human ovarian cancer.
Sigma-Aldrich
Pentostatin, ≥95% (HPLC)