Merck
  • Down-expression of GOLM1 enhances the chemo-sensitivity of cervical cancer to methotrexate through modulation of the MMP13/EMT axis.
Down-expression of GOLM1 enhances the chemo-sensitivity of cervical cancer to methotrexate through modulation of the MMP13/EMT axis.
American journal of cancer research (2018-07-24)
Rui Min Li, Man Man Nai, She Jiao Duan, Shu Xing Li, Bao Na Yin, Fang An, Yao Qing Zhai, Jie Liu, Yan Rong Chu, Yang Yu, Wen Yue Song
ABSTRACT
The highly refractory nature of cervical cancer to chemotherapeutic drugs and its epithelial-to-mesenchymal transition (EMT) are the key reasons contributing to the poor prognosis of this disease. Golgi Membrane Protein 1 (GOLM1), a protein involved in the trafficking of proteins through the Golgi apparatus, has been shown to be oncogenic in a variety of human cancers. Herein, we found GOLM1 was markedly up-regulated in cervical cancer and GOLM1 down-expression enhanced the anti-tumor effect of methotrexate. By performing mechanistic studies using both in vitro and in vivo models, we found that GOLM1 could target matrix metallopeptidase 13 (MMP13), a member of the MMPs, and regulate the EMT process. Moreover, altered EMT progression compromised the chemotherapy-enhancing effects of GOLM1 knock-down. Finally, we found significantly higher levels of GOLM1 and MMP13 in cervical cancer tissues compared with adjacent noncancerous tissues, and this was also associated with poor cervical cancer patients' prognosis. Taken together, our results suggest that the GOLM1/MMP13/EMT axis is an important factor involved in regulating methotrexate in cervical cancer, and highlights the potential of novel GOLM1-based clinical modalities as a therapeutic approach in cervical cancer patients.
Sigma-Aldrich
MISSION® esiRNA, targeting human MMP13