Merck
  • Improved sensitivity and limit-of-detection of lateral flow devices using spatial constrictions of the flow-path.
Improved sensitivity and limit-of-detection of lateral flow devices using spatial constrictions of the flow-path.
Biosensors & bioelectronics (2018-05-09)
Ioannis N Katis, Peijun J W He, Robert W Eason, Collin L Sones
ABSTRACT
We report on the use of a laser-direct write (LDW) technique that allows the fabrication of lateral flow devices with enhanced sensitivity and limit of detection. This manufacturing technique comprises the dispensing of a liquid photopolymer at specific regions of a nitrocellulose membrane and its subsequent photopolymerisation to create impermeable walls inside the volume of the membrane. These polymerised structures are intentionally designed to create fluidic channels which are constricted over a specific length that spans the test zone within which the sample interacts with pre-deposited reagents. Experiments were conducted to show how these constrictions alter the fluid flow rate and the test zone area within the constricted channel geometries. The slower flow rate and smaller test zone area result in the increased sensitivity and lowered limit of detection for these devices. We have quantified these via the improved performance of a C-Reactive Protein (CRP) sandwich assay on our lateral flow devices with constricted flow paths which demonstrate an improvement in its sensitivity by 62x and in its limit of detection by 30x when compared to a standard lateral flow CRP device.
Sigma-Aldrich
C Reactive Protein human, recombinant, expressed in E. coli, liquid
Sigma-Aldrich
Phosphate buffered saline, powder, pH 7.4, for preparing 1 L solutions
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, essentially IgG-free, low endotoxin, BioReagent, suitable for cell culture