Merck
  • Aging-Resistant Functionalized LDH⁻SAS/Nitrile-Butadiene Rubber Composites: Preparation and Study of Aging Kinetics/Anti-Aging Mechanism.
Aging-Resistant Functionalized LDH⁻SAS/Nitrile-Butadiene Rubber Composites: Preparation and Study of Aging Kinetics/Anti-Aging Mechanism.
Materials (Basel, Switzerland) (2018-05-23)
Tianxiang Li, Zhengren Shi, Xianru He, Ping Jiang, Xiaobin Lu, Rui Zhang, Xin Wang
ABSTRACT
With the aim of improving the anti-aging properties of nitrile-butadiene rubber (NBR), a functional organic filler, namely LDH⁻SAS, prepared by intercalating 4-amino-benzenesulfonic acid monosodium salt (SAS) into layered double hydroxides (LDHs) through anion exchange, was added to nitrile-butadiene rubber (NBR), giving the NBR/LDH⁻SAS composites. Successful preparation of LDH⁻SAS was confirmed by XRD, TGA and FTIR. LDH⁻SAS was well dispersed in the NBR matrix, owing to its strong interaction with the nitrile group of NBR. The obtained NBR/LDH⁻SAS composites exhibited excellent thermo-oxidative aging resistance as shown by TGA-DSC. Further investigation by ATR-FTIR indicated that SAS can capture the radical groups, even during the aging process, which largely accounts for the improved aging resistance.
Sigma-Aldrich
N-Phenyl-2-naphthylamine, 97%