Merck
  • Local lung hypoxia determines epithelial fate decisions during alveolar regeneration.
Local lung hypoxia determines epithelial fate decisions during alveolar regeneration.
Nature cell biology (2017-07-25)
Ying Xi, Thomas Kim, Alexis N Brumwell, Ian H Driver, Ying Wei, Victor Tan, Julia R Jackson, Jianming Xu, Dong-Kee Lee, Jeffrey E Gotts, Michael A Matthay, John M Shannon, Harold A Chapman, Andrew E Vaughan
ABSTRACT
After influenza infection, lineage-negative epithelial progenitors (LNEPs) exhibit a binary response to reconstitute epithelial barriers: activating a Notch-dependent ΔNp63/cytokeratin 5 (Krt5) remodelling program or differentiating into alveolar type II cells (AEC2s). Here we show that local lung hypoxia, through hypoxia-inducible factor (HIF1α), drives Notch signalling and Krt5pos basal-like cell expansion. Single-cell transcriptional profiling of human AEC2s from fibrotic lungs revealed a hypoxic subpopulation with activated Notch, suppressed surfactant protein C (SPC), and transdifferentiation toward a Krt5pos basal-like state. Activated murine Krt5pos LNEPs and diseased human AEC2s upregulate strikingly similar core pathways underlying migration and squamous metaplasia. While robust, HIF1α-driven metaplasia is ultimately inferior to AEC2 reconstitution in restoring normal lung function. HIF1α deletion or enhanced Wnt/β-catenin activity in Sox2pos LNEPs blocks Notch and Krt5 activation, instead promoting rapid AEC2 differentiation and migration and improving the quality of alveolar repair.
Sigma-Aldrich
ALW-II-41-27, ≥98% (HPLC)