Merck
  • RG7386, a Novel Tetravalent FAP-DR5 Antibody, Effectively Triggers FAP-Dependent, Avidity-Driven DR5 Hyperclustering and Tumor Cell Apoptosis.
RG7386, a Novel Tetravalent FAP-DR5 Antibody, Effectively Triggers FAP-Dependent, Avidity-Driven DR5 Hyperclustering and Tumor Cell Apoptosis.
Molecular cancer therapeutics (2016-04-03)
Peter Brünker, Katharina Wartha, Thomas Friess, Sandra Grau-Richards, Inja Waldhauer, Claudia Ferrara Koller, Barbara Weiser, Meher Majety, Valeria Runza, Huifeng Niu, Kathryn Packman, Ningping Feng, Sherif Daouti, Ralf J Hosse, Ekkehard Mössner, Thomas G Weber, Frank Herting, Werner Scheuer, Hadassah Sade, Cuiying Shao, Bin Liu, Peng Wang, Gary Xu, Suzana Vega-Harring, Christian Klein, Klaus Bosslet, Pablo Umaña
ABSTRACT
Dysregulated cellular apoptosis and resistance to cell death are hallmarks of neoplastic initiation and disease progression. Therefore, the development of agents that overcome apoptosis dysregulation in tumor cells is an attractive therapeutic approach. Activation of the extrinsic apoptotic pathway is strongly dependent on death receptor (DR) hyperclustering on the cell surface. However, strategies to activate DR5 or DR4 through agonistic antibodies have had only limited clinical success. To pursue an alternative approach for tumor-targeted induction of apoptosis, we engineered a bispecific antibody (BsAb), which simultaneously targets fibroblast-activation protein (FAP) on cancer-associated fibroblasts in tumor stroma and DR5 on tumor cells. We hypothesized that bivalent binding to both FAP and DR5 leads to avidity-driven hyperclustering of DR5 and subsequently strong induction of apoptosis in tumor cells but not in normal cells. Here, we show that RG7386, an optimized FAP-DR5 BsAb, triggers potent tumor cell apoptosis in vitro and in vivo in preclinical tumor models with FAP-positive stroma. RG7386 antitumor efficacy was strictly FAP dependent, was independent of FcR cross-linking, and was superior to conventional DR5 antibodies. In combination with irinotecan or doxorubicin, FAP-DR5 treatment resulted in substantial tumor regression in patient-derived xenograft models. FAP-DR5 also demonstrated single-agent activity against FAP-expressing malignant cells, due to cross-binding of FAP and DR5 across tumor cells. Taken together, these data demonstrate that RG7386, a novel and potent antitumor agent in both mono- and combination therapies, overcomes limitations of previous DR5 antibodies and represents a promising approach to conquer tumor-associated resistance to apoptosis. Mol Cancer Ther; 15(5); 946-57. ©2016 AACR.