Merck
  • Efficient and stable solution-processed planar perovskite solar cells via contact passivation.
Efficient and stable solution-processed planar perovskite solar cells via contact passivation.
Science (New York, N.Y.) (2017-02-06)
Hairen Tan, Ankit Jain, Oleksandr Voznyy, Xinzheng Lan, F Pelayo García de Arquer, James Z Fan, Rafael Quintero-Bermudez, Mingjian Yuan, Bo Zhang, Yicheng Zhao, Fengjia Fan, Peicheng Li, Li Na Quan, Yongbiao Zhao, Zheng-Hong Lu, Zhenyu Yang, Sjoerd Hoogland, Edward H Sargent
ABSTRACT
Planar perovskite solar cells (PSCs) made entirely via solution processing at low temperatures (<150°C) offer promise for simple manufacturing, compatibility with flexible substrates, and perovskite-based tandem devices. However, these PSCs require an electron-selective layer that performs well with similar processing. We report a contact-passivation strategy using chlorine-capped TiO2 colloidal nanocrystal film that mitigates interfacial recombination and improves interface binding in low-temperature planar solar cells. We fabricated solar cells with certified efficiencies of 20.1 and 19.5% for active areas of 0.049 and 1.1 square centimeters, respectively, achieved via low-temperature solution processing. Solar cells with efficiency greater than 20% retained 90% (97% after dark recovery) of their initial performance after 500 hours of continuous room-temperature operation at their maximum power point under 1-sun illumination (where 1 sun is defined as the standard illumination at AM1.5, or 1 kilowatt/square meter).
Sigma-Aldrich
SHT-263S Solarpur®
Sigma-Aldrich
SHT-263 Solarpur®