Skip to Content
Merck
Zolpidem ≥98% (HPLC), solid

Zolpidem

Zolpidem ≥98% (HPLC), solid
Synonym(s):
N,N,6-Trimethyl-2-(4-methylphenyl)-imidazo[1,2-a]pyridine-3-acetamide
Empirical Formula (Hill Notation):
C19H21N3O
CAS Number:
Molecular Weight:
307.39

Products

Product Category
Brand
Color
Formula Weight
Gene Alias
Physical Form
Purity
Research Area
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing