Skip to Content
Products
Cart
0
MY
EN
Products
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Login
Order Lookup
Quick Order
Cart
0
Back
Valencene
Synonym(s):
(+)-Valencene, (3
R
,4
aS
,5
R
)-4
a
,5-Dimethyl-3-isopropenyl-1,2,3,4,4
a
,5,6,7-octahydronaphthalene
Empirical Formula (Hill Notation):
C
15
H
24
CAS Number:
4630-07-3
Molecular Weight:
204.35
EC Number:
225-047-6
Products
Product Category
aroma chemicals (1)
Brand
Sigma-Aldrich (1)
Sustainability
Biological Source
Boiling Point (°C)
Feature
Formula Weight
Agency
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Special Grade
Halal (1)
Kosher (1)
Natural (1)
Filter Products
Showing 1-1 of 1
Compare
Product Number
Description
Pricing
W344303
Valencene,
natural, ≥65%
Expand
View Pricing
Choose up to 4 products to compare
Compare
Clear All
An unknown error has occured.
English - EN
Learn More