Skip to Content
Merck
Tyramine Vetec™, reagent grade, 99%

Tyramine

Tyramine Vetec™, reagent grade, 99%
Synonym(s):
2-(4-Hydroxyphenyl)ethylamine, 4-(2-Aminoethyl)phenol, 4-Hydroxyphenethylamine
Linear Formula:
HOC6H4CH2CH2NH2
CAS Number:
Molecular Weight:
137.18
EC Number:

Products

Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Gene Alias
Melting Point (°C)
Product Line
Purity
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Reference Material Type
Special Grade
Technique
Showing 1-4 of 4
Compare
Product Number
Description
Pricing