Skip to Content
Merck
TSKgel® SuperAW Guard Column phase hydroxylated methacrylate, L × I.D. 3.5 cm × 4.6 mm, 7 μm particle size

TSKgel® SuperAW (7 µm) HPLC Columns

TSKgel® SuperAW Guard Column phase hydroxylated methacrylate, L × I.D. 3.5 cm × 4.6 mm, 7 μm particle size
Synonym(s):
TSKgel® Size Exclusion (PW-Type) HPLC Guard Column

Products

Product Category
Manufacturer
Product Line
Agency
USP Suitability
Separation Technique
ID (mm)
Length (cm)
Particle Size (µm)
Technique
Showing 1-1 of 1