Skip to Content
Merck
TSKgel® CM-STAT HPLC Column phase carboxymethyl, L × I.D. 10 cm × 4.6 mm, 7 μm particle size

TSKgel® CM-STAT (7 µm) HPLC Columns

TSKgel® CM-STAT HPLC Column phase carboxymethyl, L × I.D. 10 cm × 4.6 mm, 7 μm particle size
Synonym(s):
TSKgel® CM-STAT Columns

Products

Product Category
Manufacturer
Product Line
Separation Technique
ID (mm)
Length (cm)
Particle Size (µm)
Technique
Showing 1-1 of 1