
tri-Sodium phosphate dodecahydrate

Synonym(s):
tri-Sodium phosphate dodecahydrate
Empirical Formula (Hill Notation):
Na3O4P · 12H2O
CAS Number:
Molecular Weight:
380.12
Products
Product Category
Brand
Formula Weight
Physical Form
Product Line
Agency
Quality Segment
Application
Showing 1-1 of 1
Compare
Product Number
Description
Pricing