![Trimethyltetradecylammonium bromide European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/261/649/a42d6600-58fa-4482-9b68-b15d870b38a7/640/a42d6600-58fa-4482-9b68-b15d870b38a7.png)
Trimethyltetradecylammonium bromide
![Trimethyltetradecylammonium bromide European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/261/649/a42d6600-58fa-4482-9b68-b15d870b38a7/640/a42d6600-58fa-4482-9b68-b15d870b38a7.png)
Synonym(s):
Myristyltrimethylammonium bromide, Tetradecyltrimethylammonium bromide, Trimethyl(tetradecyl)ammonium bromide
Linear Formula:
CH3(CH2)13N(Br)(CH3)3
CAS Number:
Molecular Weight:
336.39
Products
Formula Weight
Manufacturer
Melting Point (°C)
Format
Application
Markush Class
Markush Group
API Family
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing