Skip to Content
Merck
Triethylene glycol dimethacrylate contains 200 ppm monomethyl ether hydroquinone as inhibitor, 99%

Triethylene glycol dimethacrylate

Triethylene glycol dimethacrylate contains 200 ppm monomethyl ether hydroquinone as inhibitor, 99%
Linear Formula:
CH2=C(CH3)COO(CH2CH2O)3COC(CH3)=CH2
CAS Number:
Molecular Weight:
286.32
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Functionality
Shape
Polymer Composition
Polymer Type
Reaction Type
Reagent Type