Toluidine blue O (C.I.52040)
Synonym(s):
Toluidine blue O (C.I.52040)
Empirical Formula (Hill Notation):
C15H16ClN3S
CAS Number:
Molecular Weight:
305.83
Products
Brand
Formula Weight
Physical Form
Chemical Composition
Quality Segment
Application
Showing 1-1 of 1
Compare
Product Number
Description
Pricing