
Thymidine Phosphorylase, recombinant from Escherichia coli

Synonym(s):
Thymidine:orthophosphate deoxy-D-ribosyltransferase
CAS Number:
Products
Product Category
Brand
Color
Physical Form
Recombinant Host
Quality Segment
Application
Technique
Showing 1-1 of 1