Skip to Content
Merck
Sulfur LR, ≥99%

Sulfur

Sulfur LR, ≥99%
Synonym(s):
Sulfur
Empirical Formula (Hill Notation):
S
CAS Number:
Molecular Weight:
32.07
EC Number:

Products

Product Category
Brand
Sustainability
Boiling Point (°C)
Feature
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Quality Segment
Application
Reference Material Type
Special Grade
Technique