Skip to Content
Merck
(S)-(−)-β-Citronellol 99%, FG

(S)-(−)-β-Citronellol

(S)-(−)-β-Citronellol 99%, FG
Synonym(s):
β-Rhodinol, (S)-3,7-Dimethyl-6-octen-1-ol
Linear Formula:
(CH3)2C=CHCH2CH2CH(CH3)CH2CH2OH
CAS Number:
Molecular Weight:
156.27
EC Number:

Products

Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Agency
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing