
Rupatadine fumarate

Synonym(s):
8-Chloro-11-{1-[(5-methyl-3-pyridinyl)methyl]-4-piperidinylidene}-6,11-dihydro-5H-benzo[5,6]cyclohepta[1,2-b]pyridine (E)-2-butenedioate (1:1)
Empirical Formula (Hill Notation):
C26H26ClN3 · C4H4O4
CAS Number:
Molecular Weight:
532.03
Products
Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Research Area
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing