Skip to Content
Merck
Quinaldine analytical standard, ≥90% (GC)

Quinaldine

Quinaldine analytical standard, ≥90% (GC)
Synonym(s):
2-Methylquinoline
Empirical Formula (Hill Notation):
C10H9N
CAS Number:
Molecular Weight:
143.19
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Gene Alias
Melting Point (°C)
Physical Form
Purity
Format
Quality Segment
Application
Markush Class
Markush Group
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing