Skip to Content
Merck
Quercetin 3-gentiobioside phyproof® Reference Substance

Quercetin 3-gentiobioside

Quercetin 3-gentiobioside phyproof® Reference Substance
Synonym(s):
Quercetin 3-O-β-gentiobioside, 3,3′,4′,5,7-Pentahydroxyflavone 3-gentiobioside
Empirical Formula (Hill Notation):
C27H30O17
CAS Number:
Molecular Weight:
626.52

Products

Formula Weight
Manufacturer
Product Line
Format
Application
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing