
Propylene oxide

Synonym(s):
(±)-Propylene oxide, (±)-Methyloxirane, 1,2-Epoxypropane
Empirical Formula (Hill Notation):
C3H6O
CAS Number:
Molecular Weight:
58.08
Products
Boiling Point (°C)
Formula Weight
Manufacturer
Melting Point (°C)
Format
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing