Skip to Content
Merck
Phenyl salicylate ≥99%, FG

Phenyl salicylate

Phenyl salicylate ≥99%, FG
Synonym(s):
Salol
Linear Formula:
2-(HO)C6H4CO2C6H5
CAS Number:
Molecular Weight:
214.22
EC Number:

Products

Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Agency
Application
Markush Class
Markush Group
Organoleptic Class
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing