Pentylbenzene
Synonym(s):
1-Phenylpentane
Linear Formula:
C6H5(CH2)4CH3
CAS Number:
Molecular Weight:
148.24
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Manufacturer
Melting Point (°C)
Product Line
Purity
Quality Segment
Markush Class
Markush Group
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing