Parathion solution
Synonym(s):
Parathion-ethyl solution
Empirical Formula (Hill Notation):
C10H14NO5PS
CAS Number:
Molecular Weight:
291.26
Products
Product Category
Brand
Formula Weight
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing