
NPPOC-dG(ipac) RayDite™

Synonym(s):
NPPOC-dG(ipac) Amidite, NPPOC-dG(ipac) Phosphoramidite
Empirical Formula (Hill Notation):
C40H51N8O11P
Molecular Weight:
850.85
Products
Brand
Formula Weight
Product Line
Purity
Showing 1-1 of 1
Compare
Product Number
Description
Pricing