![N-Nitrosopyrrolidine certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland](/deepweb/assets/sigmaaldrich/product/structures/222/494/4d2dc1ba-14bf-44bd-97a4-ca68cb2ef44a/640/4d2dc1ba-14bf-44bd-97a4-ca68cb2ef44a.png)
N-Nitrosopyrrolidine
![N-Nitrosopyrrolidine certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland](/deepweb/assets/sigmaaldrich/product/structures/222/494/4d2dc1ba-14bf-44bd-97a4-ca68cb2ef44a/640/4d2dc1ba-14bf-44bd-97a4-ca68cb2ef44a.png)
Synonym(s):
1-Nitrosopyrrolidine, N-Nitrosopyrrolidine, NPYR
Empirical Formula (Hill Notation):
C4H8N2O
CAS Number:
Molecular Weight:
100.12
Products
Brand
Boiling Point (°C)
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2