Methenamine mandelate
Synonym(s):
Hexamethylenetetramine mandelate salt, 1,3,5,7-Tetraazatricyclo[3.3.1.13,7]decane, Hexamine, Methenamine
Empirical Formula (Hill Notation):
C6H12N4 · C8H8O3
CAS Number:
Molecular Weight:
292.33
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing