Skip to Content
Merck
Luria Broth bottle capacity × filling volume 250 mL × 200 mL, closure type, screw cap with 3 loci

Luria Broth

Luria Broth bottle capacity × filling volume 250 mL × 200 mL, closure type, screw cap with 3 loci
Synonym(s):
LB Broth

Products

Product Category
Brand
Physical Form
Agency
Capacity (mL)
Closure Type
Application
Microorganism Suitability
Package Size
Sterility
Sterilization Method
Technique
Showing 1-1 of 1