![Kaempferol 3-rutinoside phyproof® Reference Substance](/deepweb/assets/sigmaaldrich/product/structures/826/305/9d27e950-67e8-4ed9-9ceb-947d82a8a376/640/9d27e950-67e8-4ed9-9ceb-947d82a8a376.png)
Kaempferol 3-rutinoside
![Kaempferol 3-rutinoside phyproof® Reference Substance](/deepweb/assets/sigmaaldrich/product/structures/826/305/9d27e950-67e8-4ed9-9ceb-947d82a8a376/640/9d27e950-67e8-4ed9-9ceb-947d82a8a376.png)
Synonym(s):
Kaempferol 3-O-β -rutinoside, 3,4′,5,7-Tetrahydroxyflavone 3-rhamnoglucoside, Nicotiflorin
Empirical Formula (Hill Notation):
C27H30O15
CAS Number:
Molecular Weight:
594.52
Products
Formula Weight
Manufacturer
Product Line
Format
Application
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing