Skip to Content
Merck
Isopropyl β-D-thiogalactoside Vetec™, reagent grade, ≥99%

Isopropyl-β-D-thiogalactoside

Isopropyl β-D-thiogalactoside Vetec™, reagent grade, ≥99%
Synonym(s):
Isopropyl β-D-1-thiogalactopyranoside, IPTG, Isopropyl β-D-thiogalactoside
Empirical Formula (Hill Notation):
C9H18O5S
CAS Number:
Molecular Weight:
238.30

Products

Formula Weight
Manufacturer
Physical Form
Product Line
Markush Class
Markush Group