![Isopropalin certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland](/deepweb/assets/sigmaaldrich/product/structures/619/244/d21529af-5a4a-47ca-9912-220cdfaf6268/640/d21529af-5a4a-47ca-9912-220cdfaf6268.png)
Isopropalin
![Isopropalin certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland](/deepweb/assets/sigmaaldrich/product/structures/619/244/d21529af-5a4a-47ca-9912-220cdfaf6268/640/d21529af-5a4a-47ca-9912-220cdfaf6268.png)
Synonym(s):
4-Isopropyl-2,6-dinitro-N,N-dipropylaniline
Empirical Formula (Hill Notation):
C15H23N3O4
CAS Number:
Molecular Weight:
309.36
Products
Brand
Formula Weight
Manufacturer
Product Line
Quality Segment
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing