![Isoflupredone Acetate certified reference material, pharmaceutical secondary standard](/deepweb/assets/sigmaaldrich/product/images/233/674/0bdd9743-c699-479c-854d-be1b19bad9ed/640/0bdd9743-c699-479c-854d-be1b19bad9ed.jpg)
Isoflupredone acetate
![Isoflupredone Acetate certified reference material, pharmaceutical secondary standard](/deepweb/assets/sigmaaldrich/product/images/233/674/0bdd9743-c699-479c-854d-be1b19bad9ed/640/0bdd9743-c699-479c-854d-be1b19bad9ed.jpg)
Synonym(s):
9α-Fluoroprednisolone 21-acetate, (11β)-21-(Acetyloxy)-9-fluoro-11,17-dihydroxypregna-1,4-diene-3,20-dione
Empirical Formula (Hill Notation):
C23H29FO6
CAS Number:
Molecular Weight:
420.47
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2